8-Bromopyrido[3,4-b]pyrazin-5(6H)-one 97%
Supplier: Ambeed
|
Warning
|
A337919-1G
A337919-250MG
77142-030EA
394.66
USD
77142-030
77142-032
8-Bromopyrido[3,4-b]pyrazin-5(6H)-one 97%
8-Bromopyrido[3,4-b]pyrazin-5(6H)-one
Precaución: for Research Use Only
Formula:
C₇H₄BrN₃O MW: 226.03 g/mol |
CAS Number:
1590409-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...