2-Bromo-4-chlorotoluene 98%

Supplier: Ambeed
Warning

A337346-100G A337346-10G A337346-25G A337346-5G A337346-500G
77142-016EA 56.64 USD
77142-016 77142-018 77142-020 77142-022 77685-965
2-Bromo-4-chlorotoluene 98%
2-Bromo-4-chlorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MDL Number: MFCD00060650
CAS Number: 27139-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR