4-Amino-2,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A334623-10G A334623-250MG A334623-1G
77141-978EA 108.15 USD
77141-978 77141-980 77685-955
4-Amino-2,5-difluorobenzoic acid 97%
4-Amino-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD06208208
CAS Number: 773108-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR