t-Boc-N-Amido-PEG3-CH2CO2H 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azahexadecan-16-oic acid

A334552-100MG A334552-1G A334552-250MG A334552-25G A334552-5G
77141-968EA 13.39 USD
77141-968 77141-970 77141-972 77141-974 77141-976
t-Boc-N-Amido-PEG3-CH2CO2H 97%
t-Boc-N-Amido-PEG3-CH2CO2H

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₇
MDL Number: MFCD22376773
CAS Number: 462100-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR