1,3-Diisopropylimidazolium tetrafluoroborate 96%
Supplier: Ambeed
|
Warning
|
A330120-100MG
A330120-1G
A330120-250MG
A330120-5G
77141-938EA
5.15
USD
77141-938
77141-940
77141-942
77685-946
1,3-Diisopropylimidazolium tetrafluoroborate 96%
1,3-Diisopropylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₉H₁₇BF₄N₂ |
MDL Number:
MFCD08276804 CAS Number: 286014-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...