5-Chloro-2,4-difluoroaniline 97%

Supplier: Ambeed
Warning

A328894-100G A328894-10G A328894-1G A328894-25G A328894-5G
77141-920EA 405.27 USD
77141-920 77141-922 77141-924 77141-926 77141-928
5-Chloro-2,4-difluoroaniline 97%
5-Chloro-2,4-difluoroaniline

Precaución: for Research Use Only

Formula: C6H4ClF2N
MW: 163.5525 g/mol
MDL Number: MFCD06808806
CAS Number: 348-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR