3-([1,1'-Biphenyl]-4-yl)propiolic acid 95%

Supplier: Ambeed
Warning

A328582-100MG A328582-1G A328582-250MG A328582-50MG
77141-912EA 49.87 USD
77141-912 77141-914 77141-916 77141-918
3-([1,1'-Biphenyl]-4-yl)propiolic acid 95%
3-([1,1'-Biphenyl]-4-yl)propiolic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₂
MW: 222.23 g/mol
MDL Number: MFCD21013408
CAS Number: 32340-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR