(R)-4-(tert-Butoxycarbonyl)-6,6-dimethylmorpholine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A327533-100MG
A327533-250MG
77141-902EA
80.34
USD
77141-902
77141-904
(R)-4-(tert-Butoxycarbonyl)-6,6-dimethylmorpholine-2-carboxylic acid 98%
(R)-4-(tert-Butoxycarbonyl)-6,6-dimethylmorpholine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₅ |
MDL Number:
MFCD22690355 CAS Number: 1416445-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...