3,4-Difluoro-2-methylbenzonitrile 98%
Supplier: Ambeed
|
Danger
|
A326055-10G
A326055-25G
77141-896EA
231.28
USD
77141-896
77141-898
3,4-Difluoro-2-methylbenzonitrile 98%
3,4-Difluoro-2-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅F₂N MW: 153.13 g/mol |
MDL Number:
MFCD07777154 CAS Number: 847502-83-4 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...