1-Butyl-1-methylpyrrolidiniumhexafluorophosphate 99+%

Supplier: Ambeed
Warning

Synonyms: N-Butyl-N-methylpyrrolidinium hexafluorophosphate

A325616-100G A325616-1G A325616-25G A325616-5G
77141-888EA 135.96 USD
77141-888 77141-890 77141-892 77141-894
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate 99+%
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate

Precaución: for Research Use Only

Formula: C₉H₂₀F₆NP
MDL Number: MFCD08458501
CAS Number: 330671-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR