(2,2′-Bipyridine)dichloropalladium(II) 98%

Supplier: Ambeed
Warning

A324713-100MG A324713-1G A324713-250MG A324713-5G
77141-868EA 28.65 USD
77141-868 77141-870 77141-872 77141-874
(2,2′-Bipyridine)dichloropalladium(II) 98%
(2,2′-Bipyridine)dichloropalladium(II)

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂N₂Pd
MW: 333.50 g/mol
MDL Number: MFCD00134305
CAS Number: 14871-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR