(2-Carboxyethyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

A324272-100G A324272-10G A324272-1G A324272-25G A324272-500G A324272-5G
77141-856EA 74.17 USD
77141-856 77141-858 77141-860 77141-862 77141-864 77141-866
(2-Carboxyethyl)triphenylphosphonium bromide 97%
(2-Carboxyethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₀BrO₂P
MW: 415.2601 g/mol
MDL Number: MFCD00031698
CAS Number: 51114-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR