2-Bromo-1-(3-chloro-2-fluorophenyl)ethanone 97%

Supplier: Ambeed
Danger

A322654-100MG A322654-250MG
77141-842EA 44.56 USD
77141-842 77141-844
2-Bromo-1-(3-chloro-2-fluorophenyl)ethanone 97%
2-Bromo-1-(3-chloro-2-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrClFO
MW: 251.48 g/mol
MDL Number: MFCD13194234
CAS Number: 1192313-64-6
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR