1-(6-Bromo-1H-indazol-1-yl)ethanone 97%

Supplier: Ambeed
Warning

A322232-100MG A322232-1G A322232-250MG
77141-826EA 46.68 USD
77141-826 77141-828 77141-830
1-(6-Bromo-1H-indazol-1-yl)ethanone 97%
1-(6-Bromo-1H-indazol-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MW: 239.06 g/mol
MDL Number: MFCD14560558
CAS Number: 651780-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR