Ethyl 2-(1H-imidazol-4-yl)acetate 97%

Supplier: Ambeed
Warning

A321949-100MG A321949-250MG A321949-1G
77141-822EA 50.93 USD
77141-822 77141-824 77568-650
Ethyl 2-(1H-imidazol-4-yl)acetate 97%
Ethyl 2-(1H-imidazol-4-yl)acetate

Precaución: For research use only.

Formula: C₇H₁₀N₂O₂
MW: 154.16 g/mol
MDL Number: MFCD18414457
CAS Number: 28782-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR