4-(Difluoromethoxy)-3-fluorophenol 95%

Supplier: Ambeed
Warning

A320914-100MG A320914-1G A320914-250MG
77141-800EA 47.38 USD
77141-800 77141-802 77141-804
4-(Difluoromethoxy)-3-fluorophenol 95%
4-(Difluoromethoxy)-3-fluorophenol

Precaución: for Research Use Only

Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
MDL Number: MFCD18395029
CAS Number: 184162-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR