4-Hydroxy-3,5-diiodobenzaldehyde 98%

Supplier: Ambeed
Warning

A320699-1G A320699-25G A320699-5G
77141-796EA 9.26 USD
77141-796 77141-798 77685-918
4-Hydroxy-3,5-diiodobenzaldehyde 98%
4-Hydroxy-3,5-diiodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄I₂O₂
MDL Number: MFCD00014670
CAS Number: 1948-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR