2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine 95%

Supplier: Ambeed
Warning

A320127-100MG A320127-1G A320127-250MG
77141-786EA 35.01 USD
77141-786 77141-788 77141-790
2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine 95%
2-Methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₂
MDL Number: MFCD00183927
CAS Number: 1026-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR