(R)-(-)-Carvone 95%

Supplier: Ambeed
Warning

A320071-100G A320071-25G A320071-500G
77141-780EA 20.6 USD
77141-780 77141-782 77141-784
(R)-(-)-Carvone 95%
(R)-(-)-Carvone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00001578
CAS Number: 6485-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR