(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-ethoxy-5-fluorophenylboronic acid

A319528-100MG A319528-1G A319528-250MG
77141-774EA 18.55 USD
77141-774 77141-776 77141-778
(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid 97%
(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BClFO₃
MW: 218.41 g/mol
MDL Number: MFCD20441841
CAS Number: 2096334-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR