(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: CTH-(S)-P-Phos, (S)-4,4'-Bis(diphenylphosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine

A319416-100MG A319416-250MG A319416-50MG
77141-772EA 77.25 USD
77141-772 77571-906 77685-914
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥98%, ee 99%
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine

Precaución: for Research Use Only

Formula: C₃₈H₃₄N₂O₄P₂
MW: 644.64 g/mol
MDL Number: MFCD04038734
CAS Number: 362524-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR