(S)-(-)-1-(p-Tolyl)ethylamine 95%

Supplier: Ambeed
Danger

Synonyms: (S)-(-)-4-(1-Aminoethyl)toluene, (S)-1-(p-Tolyl)ethanamine

A319322-1G A319322-250MG A319322-25G A319322-5G
77141-764EA 13.8 USD
77141-764 77141-766 77141-768 77141-770
(S)-(-)-1-(p-Tolyl)ethylamine 95%
(S)-(-)-1-(p-Tolyl)ethylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00145246
CAS Number: 27298-98-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR