2,4,6-Triisopropylbenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4,6-Triisopropylbenzene-1-sulphonyl chloride

A317810-10G A317810-25G A317810-5G A317810-100G
77141-750EA 9.27 USD
77141-750 77141-752 77141-754 77685-901
2,4,6-Triisopropylbenzenesulphonyl chloride 98%
2,4,6-Triisopropylbenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₅H₂₃ClO₂S
MDL Number: MFCD00007433
CAS Number: 6553-96-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR