Ethyl 2,3,4,9-tetrahydro-1H-carbazole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A316346-100MG
A316346-1G
A316346-250MG
77141-732EA
16.98
USD
77141-732
77141-734
77141-736
Ethyl 2,3,4,9-tetrahydro-1H-carbazole-3-carboxylate 97%
Ethyl 2,3,4,9-tetrahydro-1H-carbazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₇NO₂ MW: 243.30 g/mol |
MDL Number:
MFCD00034690 CAS Number: 26088-68-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...