(R)-Pentane-1,5-diyl bis(3-((R)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate 95%
Supplier: Ambeed
|
Danger
|
A312184-1G
A312184-250MG
77141-700EA
66.94
USD
77141-700
77141-702
(R)-Pentane-1,5-diyl bis(3-((R)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate 95%
(R)-Pentane-1,5-diyl bis(3-((R)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propanoate) dioxalate
Precaución: for Research Use Only
Formula:
C₅₅H₇₀N₂O₂₀ |
MDL Number:
MFCD11973631 CAS Number: 96687-52-4 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...