2-(Methylthio)pyrido[4,3-d]pyrimidin-5(6H)-one 97%

Supplier: Ambeed
Warning

A310645-100MG A310645-1G A310645-250MG A310645-5G
77141-668EA 42.44 USD
77141-668 77141-670 77141-672 77141-674
2-(Methylthio)pyrido[4,3-d]pyrimidin-5(6H)-one 97%
2-(Methylthio)pyrido[4,3-d]pyrimidin-5(6H)-one

Precaución: for Research Use Only

Formula: C₈H₇N₃OS
MW: 193.23 g/mol
MDL Number: MFCD14756706
CAS Number: 902576-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR