(R)-(+)-1-Phenyl-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-α-Ethylbenzyl alcohol

A309310-250MG A309310-5G A309310-100MG
77141-650EA 23.68 USD
77141-650 77141-652 77746-590
(R)-(+)-1-Phenyl-1-propanol 98%
(R)-(+)-1-Phenyl-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00064279
CAS Number: 1565-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR