(2-Chloro-6-methoxypyridin-4-yl)methanol 95%

Supplier: Ambeed
Warning

A308798-100MG A308798-250MG
77141-646EA 57.29 USD
77141-646 77141-648
(2-Chloro-6-methoxypyridin-4-yl)methanol 95%
(2-Chloro-6-methoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
MDL Number: MFCD09029989
CAS Number: 193001-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR