2,2-Dimethylsuccinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

A307641-100G A307641-10G A307641-25G A307641-500G A307641-5G
77141-634EA 69 USD
77141-634 77141-636 77141-638 77141-640 77141-642
2,2-Dimethylsuccinic acid 95%
2,2-Dimethylsuccinic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MDL Number: MFCD00004196
CAS Number: 597-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR