1-Butyl-3-methylpyridiniumtetrafluoroborate 99%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-3-methylpyridin-1-ium tetrafluoroborate

A306403-25G A306403-5G
77141-612EA 19.57 USD
77141-612 77141-614
1-Butyl-3-methylpyridiniumtetrafluoroborate 99%
1-Butyl-3-methylpyridiniumtetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₆BF₄N
MDL Number: MFCD08458499
CAS Number: 597581-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR