2-Iododibenzofuran 97%

Supplier: Ambeed
Warning

Synonyms: 2-Iododibenzo[b,d]furan

A306076-1G A306076-1MG A306076-250MG
77141-606EA 146.41 USD
77141-606 77141-608 77141-610
2-Iododibenzofuran 97%
2-Iododibenzofuran

Precaución: for Research Use Only

Formula: C₁₂H₇IO
MW: 294.09 g/mol
MDL Number: MFCD00092649
CAS Number: 5408-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR