4-Bromo-2-(trifluoromethoxy)phenylacetonitrile 97+%

Supplier: Ambeed
Warning

A304718-10G A304718-1G A304718-250MG A304718-5G
77141-594EA 197.33 USD
77141-594 77141-596 77141-598 77141-600
4-Bromo-2-(trifluoromethoxy)phenylacetonitrile 97+%
4-Bromo-2-(trifluoromethoxy)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₅BrF₃NO
MW: 280.04 g/mol
MDL Number: MFCD23703476
CAS Number: 1427502-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR