2-(2-Isopropyl-5-methylphenoxy)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Isopropyl-5-methylphenoxy)acetic acid

A304642-100MG A304642-1G A304642-250MG
77141-588EA 21.22 USD
77141-588 77141-590 77141-592
2-(2-Isopropyl-5-methylphenoxy)acetic acid 98%
2-(2-Isopropyl-5-methylphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00021757
CAS Number: 5333-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR