1-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-methylmethanamine 97%

Supplier: Ambeed
Warning

A304266-1G A304266-250MG A304266-5G
77141-582EA 133.68 USD
77141-582 77141-584 77141-586
1-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-methylmethanamine 97%
1-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₁₄H₂₁BClNO₂
MW: 281.58 g/mol
MDL Number: MFCD18730284
CAS Number: 2096333-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR