(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one 95%

Supplier: Ambeed
Warning

Synonyms: (1R,5S)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one, (1R,5S)-(+)-6,6-Dimethyl-2-oxobicyclo[3.1.1]heptane, (1R)-(+)-Nopinone

A303782-1G A303782-250MG A303782-25G A303782-5G
77141-574EA 62.6 USD
77141-574 77141-576 77141-578 77141-580
(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one 95%
(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one

Precaución: for Research Use Only

Formula: C₉H₁₄O
MW: 138.207 g/mol
MDL Number: MFCD08447116
CAS Number: 38651-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR