(5-(((tert-Butyldimethylsilyl)oxy)methyl)thiophen-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A303455-100MG A303455-1G A303455-250MG
77141-568EA 37.14 USD
77141-568 77141-570 77141-572
(5-(((tert-Butyldimethylsilyl)oxy)methyl)thiophen-3-yl)boronic acid 97%
(5-(((tert-Butyldimethylsilyl)oxy)methyl)thiophen-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₁BO₃SSi
MW: 272.24 g/mol
MDL Number: MFCD28369565
CAS Number: 2122282-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR