(8-Bromonaphthalen-1-yl)methanol 95%

Supplier: Ambeed
Warning

A301696-100MG A301696-250MG A301696-50MG
77141-554EA 137.92 USD
77141-554 77141-556 77141-558
(8-Bromonaphthalen-1-yl)methanol 95%
(8-Bromonaphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.09 g/mol
MDL Number: MFCD00099247
CAS Number: 14938-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR