4-Amino-3,5-dibromobenzoic acid 97%

Supplier: Ambeed
Warning

A300530-10G A300530-1G A300530-250MG A300530-25G A300530-5G
77141-538EA 50.46 USD
77141-538 77141-540 77141-542 77141-544 77141-546
4-Amino-3,5-dibromobenzoic acid 97%
4-Amino-3,5-dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MDL Number: MFCD00075845
CAS Number: 4123-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR