(R)-1-Benzyl 4-tert-butyl 2-cyanopiperazine-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A300476-100MG A300476-1G A300476-5G
77141-532EA 35.01 USD
77141-532 77141-534 77141-536
(R)-1-Benzyl 4-tert-butyl 2-cyanopiperazine-1,4-dicarboxylate 97%
(R)-1-Benzyl 4-tert-butyl 2-cyanopiperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₃N₃O₄
MW: 345.39 g/mol
MDL Number: MFCD22377990
CAS Number: 1374669-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR