Methyl 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate 97%

Supplier: Ambeed
Warning

A300406-100MG A300406-250MG
77141-528EA 49.87 USD
77141-528 77141-530
Methyl 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate 97%
Methyl 2-((5-bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetate

Precaución: for Research Use Only

Formula: C₁₈H₁₆BrN₃O₂S
MW: 418.31 g/mol
MDL Number: MFCD22580361
CAS Number: 878671-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR