Potassium tetraphenylborate 95%

Supplier: Ambeed
Warning

A300283-100G A300283-1G A300283-5G A300283-10G
77141-522EA 139.05 USD
77141-522 77141-524 77141-526 77685-856
Potassium tetraphenylborate 95%
Potassium tetraphenylborate

Precaución: for Research Use Only

Formula: C₂₄H₂₀BK
MDL Number: MFCD03093877
CAS Number: 3244-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR