7-Bromo-1-methylindolin-2-one 98%

Supplier: Ambeed
Warning

A297982-100MG A297982-1G A297982-250MG
77141-482EA 31.94 USD
77141-482 77141-484 77141-486
7-Bromo-1-methylindolin-2-one 98%
7-Bromo-1-methylindolin-2-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MDL Number: MFCD25542210
CAS Number: 100831-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR