2-Chloro-6-fluorobenzylamine 98%

Supplier: Ambeed
Danger

A297572-10G A297572-1G
77141-460EA 26.78 USD
77141-460 77141-462
2-Chloro-6-fluorobenzylamine 98%
2-Chloro-6-fluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇ClFN
MDL Number: MFCD00042458
CAS Number: 15205-15-9
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR