(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine 95%
Supplier: Ambeed
|
Danger
|
A297261-1G
A297261-250MG
A297261-25G
A297261-5G
77141-452EA
45.32
USD
77141-452
77141-454
77141-456
77141-458
(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine 95%
(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine
Precaución: for Research Use Only
Formula:
C₁₀H₁₃Cl₂NO₃S |
MDL Number:
MFCD00243016 CAS Number: 139628-16-3 UN: 1479 ADR: 5.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...