2,3-Difluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

A297073-100G A297073-10G A297073-1G A297073-25G A297073-5G
77141-442EA 200.52 USD
77141-442 77141-444 77141-446 77141-448 77141-450
2,3-Difluorobenzyl alcohol 97%
2,3-Difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆F₂O
MW: 144.12 g/mol
MDL Number: MFCD00010312
CAS Number: 75853-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR