4-Chloro-2-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

A296049-1G A296049-25G A296049-5G
77141-424EA 26.53 USD
77141-424 77141-426 77141-428
4-Chloro-2-nitrobenzyl alcohol 98%
4-Chloro-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD00007217
CAS Number: 22996-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR