Fmoc-cis-4-fluoro-Pro-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, Fmoc-cis-4-F-Pro-OH
A295961-100MG
A295961-1G
A295961-250MG
A295961-5G
77141-418EA
60.77
USD
77141-418
77141-420
77141-422
77685-840
Fmoc-cis-4-fluoro-Pro-OH 98%
Fmoc-cis-4-fluoro-Pro-OH
Precaución: for Research Use Only
Formula:
C₂₀H₁₈FNO₄ |
MDL Number:
MFCD07781261 CAS Number: 203866-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...