2-(2-Fluorophenyl)acetohydrazide 98%

Supplier: Ambeed
Warning

A295317-1G A295317-25G A295317-5G
77141-404EA 16.98 USD
77141-404 77141-406 77141-408
2-(2-Fluorophenyl)acetohydrazide 98%
2-(2-Fluorophenyl)acetohydrazide

Precaución: for Research Use Only

Formula: C₈H₉FN₂O
MDL Number: MFCD09729319
CAS Number: 116622-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR