Methyl 1-oxo-1,2,3,4-tetrahydroisoquinoline-5-carboxylate 97%

Supplier: Ambeed
Warning

A293928-100MG A293928-250MG A293928-1G
77141-392EA 152.77 USD
77141-392 77141-394 77616-276
Methyl 1-oxo-1,2,3,4-tetrahydroisoquinoline-5-carboxylate 97%
Methyl 1-oxo-1,2,3,4-tetrahydroisoquinoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.20 g/mol
MDL Number: MFCD18250890
CAS Number: 93258-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR