2-(5-Methylisoxazol-3-yl)but-3-yn-2-ol 97%
Supplier: Ambeed
|
Warning
|
A293711-100MG
A293711-1G
A293711-250MG
77141-386EA
105.03
USD
77141-386
77141-388
77141-390
2-(5-Methylisoxazol-3-yl)but-3-yn-2-ol 97%
2-(5-Methylisoxazol-3-yl)but-3-yn-2-ol
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD21364447 CAS Number: 1202769-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...